Preparative HPLC columns, Ultra Aqueous C18
Leverandør: Restek
9178517 9178518 9178519 9178557 9178558 9178559 9178567 9178568 9178569 9178577 9178578 9178579
REST9178517EA 34400 NOK
REST9178517 REST9178518 REST9178519 REST9178557 REST9178558 REST9178559 REST9178567 REST9178568 REST9178569 REST9178577 REST9178578 REST9178579
Preparative HPLC columns, Ultra Aqueous C18
Kromatografikolonner
Ultra Aqueous C18 is a rugged, reversed phase column with a well balanced retention profile that boasts high reproducibility and compatibility with many mobile phase, even in 100% aqueous conditions.
  • Effectively retain more types of solutes than a conventional C18
  • Ideal for multicomponent LC-MS analyses
  • When used with a gradient it eliminates multiple compounds eluting near the column void time
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 15%
Kappet ved endene N/A
pH område 2,5 - 8
Temperaturgrense 80 °C
USP kode L1