Analytical HPLC columns, Ultra Aromax (USP L11)
Leverandør: Restek
9127362 9127351 9127311 9127312 9127312-700 9127313-700 9127315 9127315-700 912731E 9127331 9127332 9127332-700 9127333-700 9127335 9127335-700 912733E 9127352 9127352-700 9127353-700 9127355 9127355-700 912735E 9127361 9127362-700 9127363-700 9127365 9127365-700 912736E 912750210 9127511 9127512 9127512-700 9127513-700 9127515 9127515-700 912751E 9127521 9127522 9127522-700 9127523-700 9127525 9127525-700 912752E 9127531 9127532 9127532-700 9127533-700 9127535 9127535-700 912753E 9127551 9127552 9127552-700 9127553-700 9127555 912750212 9127555-700 912755E 9127561 9127562 9127562-700 9127563-700 9127565 9127565-700 912756E 9127571 9127572 9127572-700 9127573-700 9127575 9127575-700 912757E
REST9127362EA 13950 NOK
REST9127362 REST9127351 REST9127311 REST9127312 REST9127312-700 REST9127313-700 REST9127315 REST9127315-700 REST912731E REST9127331 REST9127332 REST9127332-700 REST9127333-700 REST9127335 REST9127335-700 REST912733E REST9127352 REST9127352-700 REST9127353-700 REST9127355 REST9127355-700 REST912735E REST9127361 REST9127362-700 REST9127363-700 REST9127365 REST9127365-700 REST912736E REST912750210 REST9127511 REST9127512 REST9127512-700 REST9127513-700 REST9127515 REST9127515-700 REST912751E REST9127521 REST9127522 REST9127522-700 REST9127523-700 REST9127525 REST9127525-700 REST912752E REST9127531 REST9127532 REST9127532-700 REST9127533-700 REST9127535 REST9127535-700 REST912753E REST9127551 REST9127552 REST9127552-700 REST9127553-700 REST9127555 REST912750212 REST9127555-700 REST912755E REST9127561 REST9127562 REST9127562-700 REST9127563-700 REST9127565 REST9127565-700 REST912756E REST9127571 REST9127572 REST9127572-700 REST9127573-700 REST9127575 REST9127575-700 REST912757E
Analytical HPLC columns, Ultra Aromax (USP L11)
Kromatografikolonner
These Ultra Aromax is a unique reversed-phase material that exhibits extreme retention and selectivity for aromatic and/or unsaturated compounds. They are made of high-purity, type-B silica that minimises activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications.
  • It is an alternative to Biphenyl phase when increased retention is required
  • Suitable for gradient LC-MS analyses when conventional columns are not giving adequate retention or selectivity
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 17%
Kappet ved endene Ja N/A
pH område 2,5 - 8 2,5 - 8
Temperaturgrense 80 °C 80 °C
USP kode L11