Analytical HPLC columns, Ultra C1 (USP L13)
Leverandør: Restek
9101515 9101521 9101572 9101575 9101312-700 9101313-700 9101315-700 9101332-700 9101333-700 9101335-700 9101352-700 9101353-700 9101355-700 9101362-700 9101363-700 9101365-700 9101512-700 9101513-700 9101515-700 9101522-700 9101523-700 9101525-700 9101532-700 9101533-700 9101535-700 9101552-700 9101553-700 9101555-700 9101562-700 9101563-700 9101565-700 9101572-700 9101573-700 9101575-700 910131E 910133E 910135E 910136E 910151E 910152E 910153E 910155E 910156E 910157E
REST9101515EA 9025 NOK
REST9101515 REST9101521 REST9101572 REST9101575 REST9101312-700 REST9101313-700 REST9101315-700 REST9101332-700 REST9101333-700 REST9101335-700 REST9101352-700 REST9101353-700 REST9101355-700 REST9101362-700 REST9101363-700 REST9101365-700 REST9101512-700 REST9101513-700 REST9101515-700 REST9101522-700 REST9101523-700 REST9101525-700 REST9101532-700 REST9101533-700 REST9101535-700 REST9101552-700 REST9101553-700 REST9101555-700 REST9101562-700 REST9101563-700 REST9101565-700 REST9101572-700 REST9101573-700 REST9101575-700 REST910131E REST910133E REST910135E REST910136E REST910151E REST910152E REST910153E REST910155E REST910156E REST910157E
Analytical HPLC columns, Ultra C1 (USP L13)
Kromatografikolonner
Ultra C1 columns made of high-purity, type-B silica are designed for selective and reliable HPLC applications.
  • Exceptionally stable C1 phase
  • Least retentive reversed phase hydrocarbon packing
  • Minimises activity and creates high density bonding
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning 5%
Kappet ved endene Ja
pH område 2,5 - 8
Temperaturgrense 80 °C
USP kode L13