Anti-ITGAL Mouse Monoclonal Antibody (CF488A) [clone: Cris-3]

Leverandør: Biotium
BNC880151-500 BNC880151-100
BNC880151-500EA 6525 NOK
BNC880151-500 BNC880151-100
Anti-ITGAL Mouse Monoclonal Antibody (CF488A) [clone: Cris-3]
Antistoffer
Recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop II; Code N202). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: Cris-3
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR