Anti-ITGAL Mouse Monoclonal Antibody (Purified) [clone: DF1524]

Leverandør: Biotium
BNUB0312-500 BNUB0312-100
BNUB0312-500EA 5950 NOK
BNUB0312-500 BNUB0312-100
Anti-ITGAL Mouse Monoclonal Antibody (Purified) [clone: DF1524]
Antistoffer
Recognizes a protein of 180 kDa, identified as CD11a (Leucocyte Workshop IV; Code 1524). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a /C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses. This MAb potently blocks LFA-1 dependent homotypic cell aggregation.

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: DF1524
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR