Anti-SILV Mouse Monoclonal Antibody (CF568) [clone: PMEL/783]
Leverandør: Biotium
BNC680783-500 BNC680783-100
BNC680783-500EA 6875 NOK
BNC680783-500 BNC680783-100
Anti-SILV Mouse Monoclonal Antibody (CF568) [clone: PMEL/783]
Antistoffer
Cytotoxic T lymphocytes (CTL's) recognize melanoma-associated antigens, which belong to three main groups. These groups include tumor-associated testis-specific antigens, melanocyte differentiation antigens and mutated or aberrantly expressed antigens, which are routinely used as markers to identify melanomas based on their binding to specific monoclonal antibodies. gp100, also designated ME20-M, ME20-S and PMEL 17, is classified as a melanocyte differentiation antigen and is expressed at low levels in normal cell lines and tissues, but is upregulated in melanocytes. gp100 is a highlyMore Product Information