Anti-SEMA4D Mouse Monoclonal Antibody (Purified) [clone: 133-1C6]

Leverandør: Biotium
BNUB0932-500 BNUB0932-100
BNUB0932-500EA 6275 NOK
BNUB0932-500 BNUB0932-100
Anti-SEMA4D Mouse Monoclonal Antibody (Purified) [clone: 133-1C6]
Antistoffer
Recognizes a homodimeric protein comprised of 50 kDa subunits, identified as CD100 (Workshop VI; Code N-L026). It is expressed on majority of haemopoietic cells (B, T, NK and myeloid cells) and is absent from bone marrow, erythrocytes, eosinophils and endothelial cells. Its expression is increased after PHA-activation. CD100 was shown to associate with different partner molecules in T cells such as CD45, a key molecule with protein tyrosine phosphatase activity involved in T-cell transduction, and a Serine kinase. It plays a role in homotypic cell adhesion and in T cell activation.

Bestill nå

SPESIFIKASJONER

Antigensymbol SEMA4D
Antigen-navn Semaphorin 4D
Environmentally Preferable
Antigen synonymer M sema G|Semaphorin-4D|MSEMA|Semaphorin C like 2|Collapsin4|SEMA4D|Semcl2|Semaphorin J|SEMAJ|SemaH|Semacl2|Semaphorin H|COLL4|GR3
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon Clone 133-1C6
Reaktivitet Baboon,Human
Vert Mouse
Gen-ID 494406
Isotype IgM kappa
ImmunoFluorescens Yes
Konsentrasjon 0,2 mg/ml
Lagringsbuffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR