Anti-CD47 Mouse Monoclonal Antibody (CF405S) [clone: IAP/964]

Leverandør: Biotium
BNC040964-500 BNC040964-100
BNC040964-500EA 6875 NOK
BNC040964-500 BNC040964-100
Anti-CD47 Mouse Monoclonal Antibody (CF405S) [clone: IAP/964]
Antistoffer
This antibody reacts with Ig domain of CD47 protein. CD47, originally named integrin-associated protein (IAP), is a 50 kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: IAP/964
Conjugation: CF405S
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol CD47
Antigen synonymer IAP|Integrin Associated Protein|MER6|Antigenic Surface Determinant Protein OA3|Protein MER6|OA3|Rh Related Antigen|Integrin Associated Signal Transducer
Antistofftype Primary
Klonalitet Monoclonal
Konjugering CF® 405S
Klon Clone IAP/964
Reaktivitet Human,Mouse
Vert Mouse
Gen-ID 446414
Isotype IgG1
ImmunoKjemi Yes
ImmunoFluorescens Yes
Konsentrasjon 0,1 mg/ml
Lagringsbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR