Cut resistant gloves, HyFlex® 11-425
Leverandør: Ansell
11-425/10 11-425/11 11-425/6 11-425/7 11-425/8 11-425/9
ANSE11-425/10EA 3775 NOK
ANSE11-425/10 ANSE11-425/11 ANSE11-425/6 ANSE11-425/7 ANSE11-425/8 ANSE11-425/9
Cut resistant gloves, HyFlex® 11-425
Hansker
These seamless liners with INTERCEPT™ technology yarn are ³/₄ coated with water-based PU and nitrile.
  • Water and oil repellent treatment on both liner and coating
  • Silicone-free
  • Highest level of cut protection (level 5)
  • Excellent abrasion resistance
  • Water based process environmentally friendly innovative coating
  • Colour coded indicator button (red) corresponds to cut level: 1 (green), 3 (orange), 5 (red)
Ideal for handling glass sheets, sharp-edged objects and metal sheets. Also suitable for the cutting of dry, painted or galvanised pieces andMore Product Information
Bestill nå
SPESIFIKASJONER
Modell 11-425
Ambidekstral/håndspesifikk Håndspesifikk
Environmentally Preferable
Engangs/flergangs gjenbrukbar
Linermateriale PE
Overflate Palm and fingertips coated
Type mansjett Strikket håndledd
EN norm EN 388: 2003 (4543), EN 388: 2016 (4X43C)
Godkjent for kontakt med matvarer +