DNA methylation kit, blood and tissue
Leverandør: ENZO LIFE SCIENCES
ENZ450040050 ENZ450040200
ENZOENZ450040050EA 4125 NOK
ENZOENZ450040050 ENZOENZ450040200
DNA methylation kit, blood and tissue
Assay Nucleic Acid Assays
Blood and Tissue DNA Methylation Kit is a simple and reliable DNA bisulfite conversion directly from blood, tissue and cells without the prerequisite of DNA purification.
  • Compatible with small sample inputs – as few as 10 cells or 50 pg DNA
  • Suited for FFPE and LCM-derived samples with easy to follow instructions
Desulphonation and clean-up of the converted DNA is performed using a low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Leveringsinformasjon: Kit contains Proteinase K and storage buffer, Methylation digestion buffer (2X), BT conversion reagent, Methylation dilution buffer, Methylation-solubilisation buffer, Methylation reaction buffer, Methylation binding buffer, Methylation wash buffer concentrate, M-desulphonation buffer, Methylation elution buffer, IC columns and collection tubes.