DNA methylation kit, express
Leverandør: ENZO LIFE SCIENCES
ENZ450010050 ENZ450010200
ENZOENZ450010050EA 4250 NOK
ENZOENZ450010050 ENZOENZ450010200
DNA methylation kit, express
Assay Nucleic Acid Assays
Express DNA Methylation Kit can be used for rapid and reliable bisulfite treatment and conversion of DNA for methylation analysis.
  • High yield and ready to use
  • Easy to follow instructions
Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Leveringsinformasjon: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, Methylation elution buffer , IC column, IC collection tube and L-Desulphonation reagent.