Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Leverandør: Corning
3985 3643 3985BC 3540 3542 3544 3766 3762 3764 3768 3770 3764BC 3770BC 3763 3765 3767 3769 4588 4596 4690
734-1200EA 23375 NOK
734-1200 734-4118 734-4168 734-4086 734-4087 734-4088 CORN3766 CORN3762 CORN3764 CORN3768 738-0210 CORN3764BC 738-0211 CORN3763 CORN3765 CORN3767 CORN3769 CORN4588 CORN4596 CORN4690
Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Mikroplater HTS-mikroplater
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat RECs, ISO 14001 - Kennebunk