VWR® VisiLight® I-LED, LED-belysningssystem
Leverandør: VWR Collection
631-1754EA 4075 NOK
631-1754 631-1755 631-1775 631-1781 631-1271
VWR® VisiLight® I-LED, LED-belysningssystem
Lys LED-belysnings
VisiLight® I-LED incident lights, with the unique 'modul-LED®' concept, makes it easy to adapt light colour and beam angle to any application simply by exchanging the LED modules. Thanks to the flexible gooseneck arms, the light can be directed where it is needed.
  • Suitable for positioning on a stand, attach to a microscope, or fix to a wall (using optional accessories)
  • Include module(s) with 6000 K spot optic (3 W LED)
  • User friendly intensity control unit with memory function
W×D×H: 80×40×56 mm
Sertifiseringer: IP 40.
Bestillingsinformasjon: More Product Information