Grade 3 qualitative filter papers, Whatman™
Leverandør: Whatman products (Cytiva)
1003-055 1003-070 1003-090 1003-110 1003-125 1003-150 1003-185 1003-240 1003-320 1003-917 1003-323
512-1016EA 390 NOK
512-1016 512-1017 512-1018 512-1019 512-1020 512-1021 512-1022 512-0383 512-0464 512-1023 512-0465
Grade 3 qualitative filter papers, Whatman™
Filter Filterpapir Kvalitativt filterpapir
A standard grade filter used in qualitative analytical techniques to determine and identify materials. For use in büchner funnels.
  • Thickness of 6 µm
Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Double the thickness of Grade 1 with still finer particle retention and excellent loading capacity; more precipitate can be held without clogging. The extra thickness gives increased wet strength and makes this grade highly suitable for use in BüchnerMore Product Information
Bestill nå
SPESIFIKASJONER
Nr. 3
Vekt 185 g/m²
Environmentally Preferable
Tykkelse 390 µm
Partikkelretensjon 6 µm
Filtreringshastighet 325 s (Herzberg)
Askeinnhold 0,06%
Luftgjennomstrømning 26 s/100 ml/6,5 cm²