Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
Supplier: Whatman products (Cytiva)
10311447 10311451
515-5606EA 1550 NOK
515-5606 515-5607
Qualitative filter papers, standard grades, grade 593 ¹/₂, Whatman™
Filters Filter Papers Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. A standard grade filter paper for fine precipitates.
  • Increased filter surface area increases total loading capacity and decreases overall filtration time
  • Reduced contact between filter and funnel maintains flow rate
  • Eliminated need to quadrant-fold filter circles to fit conical funnels saves time in repetitive or multiple analyses
  • Prepleated filters have the same technical properties as the respective filter circles
Prepleated qualitative filtersMore Product Information
Order Now
SPECIFICATIONS
Grade 593 ½
Environmentally Preferable
Weight 85 g/m²
Thickness 170 µm
Particle retention 5 µm
Filtration speed 450 s (Herzberg)