MMP Fluorometric drug discovery kits, RED and GREEN
Leverandør: ENZO LIFE SCIENCES
BMLAK3010001 BMLAK3020001 BMLAK3030001 BMLAK3040001 BMLAK3060001 BMLAK3070001 BMLAK3110001 BMLAK3120001
ENZOBMLAK3010001EA 8775 NOK
ENZOBMLAK3010001 ENZOBMLAK3020001 ENZOBMLAK3030001 ENZOBMLAK3040001 ENZOBMLAK3060001 ENZOBMLAK3070001 ENZOBMLAK3110001 ENZOBMLAK3120001
MMP Fluorometric drug discovery kits, RED and GREEN
Assay Enzymanalyser
The matrix metalloproteinase (MMP) RED and GREEN Drug Discovery Kits are complete assay systems designed to screen MMP inhibitors, using long-wavelength quenched fluorogenic substrates OmniMMP™ RED or Fluorogenic Substrate (GREEN). The assays are performed in a convenient 96-well microplate format, with the compound NNGH also included as a prototypic control inhibitor.
  • Improved red-shifted substrate with better kinetics and brighter signal
  • Includes active recombinant enzyme, substrate, and assay buffer
  • Convenient real-time kinetics of cleavage is easily determined
  • Microplate format for high-throughput screening
Proteins of the matrix metalloproproteinase (MMP) family are collectively involved in the breakdown of extracellular matrix proteins and processing of bioactive molecules and are required for some normal processes. Unfortunately, these proteins are also associated with many pathological processes including cirrhosis, arthritis, metastasis, and chronic cardiovascular diseases. Collagenases (MMP-1, MMP-8, and MMP-13) and gelatinases (MMP-2 and MMP-9) also break down skin collagens, contributing to aging and wrinkling.
Leveringsinformasjon: Each kit contains 1 vial MMP enzyme, 1 vial substrate, 1 vial calibration standard, 1 vial control inhibitor (NNGH), 1 bottle (20 ml) assay buffer, 1 black 96-well microplate and instructions.