Green detection kit, LYSO-ID®
Leverandør: ENZO LIFE SCIENCES
ENZ510340100 ENZ51034K500
ENZOENZ510340100EA 1950 NOK
ENZOENZ510340100 ENZOENZ51034K500
Green detection kit, LYSO-ID®
Assay Cellulær analyse
LYSO-ID® Green Detection Kit is an acidic organelle-selective dye specially designed to use with RFP-expressing cell lines, as well as cells expressing blue, cyan or orange fluorescent proteins (BFPs, CFPs, OFPs).
  • Used for both short-term and long-term tracking studies
  • Low micromolar concentrations is sufficient for staining mammalian cells
  • Accumulates in acidic compartments (endosomes, lysosomes, and secretory vesicles)
  • Generates emission profiles that can be multiplexed with other fluorophores
LYSO-ID® Green dye is a new green-emitting, cell-permeable small organic probe molecule that spontaneously localizes to live cell acidic organelles. It can be readily used in combination with other common UV and visible light excitable fluorescent dyes and various fluorescent proteins in multicolor imaging and detection applications.
Leveringsinformasjon: Kit contains LYSO-ID® Green Detection Reagent, Hoechst 33342 nuclear stain, Chloroquine control and 10X assay buffer.