Aggresome detection kit, PROTEOSTAT®
Leverandør: ENZO LIFE SCIENCES
ENZ510350025 ENZ51035K100
ENZOENZ510350025EA 1950 NOK
ENZOENZ510350025 ENZOENZ51035K100
Aggresome detection kit, PROTEOSTAT®
Assay Cellulær analyse
PROTEOSTAT® Aggresome Detection Kit is a robust, quantitative detection of aggresomes relevant to the study of neurodegenerative disease, liver disease and toxicology.
  • Fixed-cell assay is optimised for antibody co-localisation studies
  • Easily quantifies aggresome and related inclusion bodies by flow cytometry
  • Useful for the study of neurodegenerative diseases, liver disease, toxicology studies and much more
  • Reliable and simple assay doesn’t require non-physiological protein mutations or genetically engineered cell lines
  • Validated under a wide range of conditions and with small molecule modulators demonstrating suitability for screening compounds of potential therapeutic value
The PROTEOSTAT® Aggresome Detection Kit is potentially applicable to the identification of small molecules that inhibit aggresome formation as well as immuno-localisation studies between aggregated protein cargo and the various proteins implicated in aggresome formation, such as histone deacetylase 6, parkin, ataxin-3, dynein motor complex and ubiquilin-1.
Leveringsinformasjon: Kit contains PROTEOSTAT® Aggresome Detection Reagent, Hoechst 33342 nuclear stain, Proteasome inhibitor (MG-132) and 10X assay buffer.