Glass microfibre filters with binder, grade GF 6, Whatman™
Leverandør: Whatman products (Cytiva)
10370002 10370003 10370004 10370005 10370006 10370007 10370008 10370010 10370011 10370012 10370018 10370019 10370020 10370050 10370043
515-0085EA 2600 NOK
515-0085 515-9102 515-9103 513-5000 515-9105 515-9106 515-9107 515-9108 515-9114 513-0068 515-9111 515-9115 515-9113 513-0101 WHAT10370043
Glass microfibre filters with binder, grade GF 6, Whatman™
Filter Glassfiberfilter
These microfiber filters made from borosilicate glass are suitable for water and wastewater analysis, with extremely fine particle retention and added strength.
  • Sub micron filter retention efficiency to capture extremely small particles
  • Inorganic binder for increased strength
  • Can be used as a prefilter for dense samples
Bestill nå
SPESIFIKASJONER
Nr. GF 6
Environmentally Preferable
Vekt 80 g/m² 80 g/m²
Tykkelse 350 µm 350 µm
Luftgjennomstrømning 40 s/100 ml/1,56 cm²
Maks. temp omgivelser 500 °C 500 °C