PLOT columns, Rt-alumina BOND/Na₂SO₄ columns (fused silica)
Leverandør: Restek
19757 19758 19756
REST19757EA 16350 NOK
REST19757 REST19758 REST19756
PLOT columns, Rt-alumina BOND/Na₂SO₄ columns (fused silica)
Kromatografikolonner
Rt-alumina BOND/Na₂SO₄ columns (fused silica PLOT) are used for Na₂SO₄ deactivation.
  • Acetylene and propadiene elute after butanes
  • Ideal separation for butene isomers (impurities in butene streams)
  • Methyl acetylene elutes after 1,3-butadiene
  • Cyclopropane (impurity in propylene) elutes well before propylene
  • Stable to 200 °C
Similar phases: GS-alumina,CP-Al₂O₃/Na₂SO₄, alumina-sulphate, AT-alumina.