Capillary columns, Rxi®-17Sil MS columns (fused silica)
Leverandør: Restek
14123 43820 43821 14126 14102
REST14123EA 15050 NOK
REST14123 REST43820 REST43821 REST14126 REST14102
Capillary columns, Rxi®-17Sil MS columns (fused silica)
Kromatografikolonner
These fused silica columns have midpolarity Crossbond® phase manufactured for column to column reproducibility suited for validated methods.
  • Inert column and highly selective for active environmental compounds such as PAHs
  • Low bleed for use with sensitive detectors such as MS
  • Equivalent to USP phase G3
Similar phases: DB-17ms, VF-17ms