HPV type 31/33/51 probe, PATHO-GENE®
Leverandør: ENZO LIFE SCIENCES
ENZ32887 ENZ328876000
ENZOENZ32887EA 9225 NOK
ENZOENZ32887 ENZOENZ328876000
HPV type 31/33/51 probe, PATHO-GENE®
Assay Immunologisk analyser
The PATHO-GENE® HPV type 6/11 probe is a mixture of biotin-labeled HPV 6 and HPV 11-specific probes in buffered formamide and hybridisation enhancers.
The PATHO-GENE® HPV type 6/11 probe employs a mixture of the HPV 31, HPV 33 and HPV 51-specific probes that detect HPV types that have been associated with either condyloma or cervical intraepithelial neoplasia (CIN) and carcinoma in situ (CIS). The probe mixture is denatured and hybridised to the tissue section fixed and pretreated on the microscope slide. Specific hybridisation between the HPV DNA probe and DNA in the specimen is determined by the detection of biotin.