Portable refractometers for salinity, PAL series
Leverandør: Atago
4404 4405 4506
635-0215EA 4150 NOK
635-0215 635-0216 ATAG4506
Portable refractometers for salinity, PAL series
Refraktometere Refraktometere, håndholdte
These portable refractometers are designed to deliver rapid results.
Sertifiseringer: IP 65 allows rinsing off the sample with running water.
Leveringsinformasjon: Supplied with two AAA batteries.