VWR®, Erlenmeyerkolbe med NS-slip
Leverandør: VWR Collection
201-1385EA 260 NOK
201-1385 201-1386 201-1387 201-1388 201-1389 201-1390 201-1391 201-1392 201-1393 201-1394 201-1395 201-1372 201-1373 201-1362 201-1363 201-1369 201-1370
VWR®, Erlenmeyerkolbe med NS-slip
Kolber Erlenmeyerkolber
Borsilikatglass, 3.3, Klar.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Good value alternative for a wide range of laboratory applications. Volume graduations and labelling area.
DIN 12387
Sertifiseringer: DIN 12387