Zeocin® 100 mg/ml in HEPES cell culture tested, sterilfiltrert
Leverandør: Thermo Fisher Scientific
Warning
J67140.XF J67140.8X
J67140.XFEA 1100 NOK
J67140.XF J67140.8X
Zeocin® 100 mg/ml in HEPES cell culture tested, sterilfiltrert
Zeocin®
Zeocin® is a selective antibiotic that acts on both eukaryotic and prokaryotic cells. Resistance to Zeocin® is conferred by the Sh ble gene from Streptoalloteichus hindustanus.
Zeocin® is the commercial name for a special formulation containing Phleomycin, a copper-chelated glycopeptide antibiotic isolated from a mutant strain of Streptomyces verticillus. This antibiotic of the bleomycin family exhibits activity against bacteria, eukaryotic microorganisms, plant and animal cells. Although bleomycin antibiotics perturb plasma membranes, their activity is generally believed to be related to their ability to bind and intercalate DNA thus destroying the integrity of the double helix.
NB: Research Use only: Not intended for animal or human diagnostic or therapeutic use. Recommended storage temperature: 2-8°C or -20°C. Zeocin is a trademark of InvivoGen.
MDL nummer: MFCD28100499
CAS-nummer: 11031-11-1
Bestill nå
Spesifikasjon Testresultater
Quality Control Each lot is thoroughly tested to ensure the absence of lot-to-lot variation.
Endotoxin level < 1 EU/mg
Physicochemical characterization HPLC, pH, appearance
Cell-culture tested potency validated in Zeocin®-sensitive and Zeocin®-resistant mammalian cell lines
Non-cytotoxicity of trace contaminants absence of long-term effects confirmed in Zeocin®-resistant cells