Immobilised polysaccharide columns
Leverandør: CHIRAL TECHNOLOGIES
80324 80325 80322 80345 80335 81325 81324 81335 81337 81345 80311 81311 80337 80435 80445 81435 81445 83311 83325 83324 83335 83337 83345 83435 83445 80511 80522 81524 83525 83524 83523 83522 83511 81594 81525 81523 81522 81511 80594 80525 80524 80523 83594 80225 80224 80226 80323 80423 81322 81323 81423 82224 82225 82226 83224 83225 83226 83322 83323 83423 84311 84322 84323 84324 84325 84335 84337 84345 84394 84423 84435 84445 84511 84522 84523 84524 84525 84594 85311 85322 85323 85324 85325 85335 85337 85345 85394 85423 85435 85445 85511 85522 85523 85524 85525 85594 86S84 81S82 86S83 86S82 85S84 85S83 85S82 84S84 84S83 84S82 83S84 83S83 83S82 81S83 80S84 80S83 80S82 81S84 87423 87435 87445 88335 80G24 88337 80G25 80G35 80G45 83G24 83G25 83G35 83G45 84G24 84G25 84G35 84G45 85G24 85G25 85G35 85G45 86G24 86G25 86G35 86G45 87G24 87G25 87G35 87G45 88224 88225 88226 88G24 88G25 88G35 88G45 88311 88322 88323 88324 88325 88345 88394 88423 88435 88445 88511 88522 88523 88524 88525 88592 88593 88594 88595 88S82 88S83 88S84 81U82 81U83 83U82 83U83 87U82 87U83 80432 88432 83432 87432 80442 88442 83442 87442 93511 93522 93523 93524 93525 93593 93594 93595 93S82 93S83 93S84
554-0105EA 34025 NOK
554-0105 554-0106 554-3430 554-0109 554-0108 554-0111 554-3435 554-3436 554-3437 554-3438 554-3760 554-3762 554-3761 DAIC80435 DAIC80445 DAIC81435 DAIC81445 554-3763 554-3764 554-3452 554-3453 554-3454 554-3455 DAIC83435 DAIC83445 554-2515 554-2516 554-3443 554-3461 554-3460 554-3459 554-3458 554-3457 554-3445 554-3444 554-3442 554-3441 554-3440 554-2520 554-2519 554-2518 554-2517 554-3462 554-4605 554-3428 554-3429 554-3431 554-3432 554-3433 554-3434 554-3439 554-3446 554-2354 554-3447 554-3448 554-3449 554-3450 554-3451 554-2521 554-3456 554-2522 554-3463 554-3464 554-2523 554-2524 554-3465 554-3466 554-3467 554-3468 554-2525 554-3469 554-3470 554-2526 554-3471 554-3472 554-3473 554-3474 554-3475 554-2527 554-3476 554-3477 554-3478 554-2528 554-3479 554-3480 554-3481 554-3482 554-3483 554-3484 554-3485 554-2529 554-3486 554-3487 554-3488 554-3489 554-3490 554-4623 554-4609 554-4622 554-4621 554-4620 554-4619 554-4618 554-4617 554-4616 554-4615 554-4614 554-4613 554-4612 554-4610 554-4608 554-4607 554-4606 554-4611 DAIC87423 DAIC87435 DAIC87445 DAIC88335 DAIC80G24 DAIC88337 DAIC80G25 DAIC80G35 DAIC80G45 DAIC83G24 DAIC83G25 DAIC83G35 DAIC83G45 DAIC84G24 DAIC84G25 DAIC84G35 DAIC84G45 DAIC85G24 DAIC85G25 DAIC85G35 DAIC85G45 DAIC86G24 DAIC86G25 DAIC86G35 DAIC86G45 DAIC87G24 DAIC87G25 DAIC87G35 DAIC87G45 DAIC88224 DAIC88225 DAIC88226 DAIC88G24 DAIC88G25 DAIC88G35 DAIC88G45 DAIC88311 DAIC88322 DAIC88323 DAIC88324 DAIC88325 DAIC88345 DAIC88394 DAIC88423 DAIC88435 DAIC88445 DAIC88511 DAIC88522 DAIC88523 DAIC88524 DAIC88525 DAIC88592 DAIC88593 DAIC88594 DAIC88595 DAIC88S82 DAIC88S83 DAIC88S84 DAIC81U82 DAIC81U83 DAIC83U82 DAIC83U83 DAIC87U82 DAIC87U83 DAIC80432 DAIC88432 DAIC83432 DAIC87432 DAIC80442 DAIC88442 DAIC83442 DAIC87442 DAIC93511 DAIC93522 DAIC93523 DAIC93524 DAIC93525 DAIC93593 DAIC93594 DAIC93595 DAIC93S82 DAIC93S83 DAIC93S84
Immobilised polysaccharide columns
Kromatografikolonner
These columns allow chromatographers to employ essentially any organic solvent as mobile phase to develop methods for the most challenging separations. These columns are available in a variety of particle sizes and offer great speed and column resolving power. Columns packed with 3 µm CSPs offer high speed separations and a reduction in analysis time and cost per sample. Columns, packed with 3 µm particles, may be used in conventional HPLC units without significant loss in performance, even with the smaller column dimensions.
The immobilised columns are used for applications ranging fromMore Product Information