Isopropyl-β-D-tiogalactopyranosid (from plant origin galactose) ≥99.5% (by HPLC) dioxane-free
Leverandør: PanReac AppliChem
Synonymer: IPTG
A7211.0025 A7211.1000
A7211.0025EA 14650 NOK
A7211.0025 A7211.1000
Isopropyl-β-D-tiogalactopyranosid (from plant origin galactose) ≥99.5% (by HPLC) dioxane-free
Isopropyl-β-D-tiogalactopyranosid
Isopropyl β-D-thiogalactopyranoside (IPTG) is derived from plant origin galactose.
  • Assay: min. 99,5%
  • Solid form
  • Dioxane free
  • Soluble in 5% water
  • Clear and colourless
  • Storage temperature: −20 °C
Assay (HPLC): min. 99.5 %
Solubility (5 %; H2O): clear, colorless
α20°C/D; 1 %, H2O: -29° - -33°
Dioxane: not detectable
Water (K.F.): max. 1.0 %
NB: Storage temperature: 2-8 °C
Formel: C₉H₁₈O₅S
MW: 238,31 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00063273
CAS-nummer: 367-93-1