Abbe refractometers, NAR
Leverandør: Atago
1220 1230 1240 1211 1212
ATAG1220EA 53700 NOK
ATAG1220 ATAG1230 ATAG1240 ATAG1211 ATAG1212
Abbe refractometers, NAR
Refraktometere Refraktometer, benkmodell
The NAR Abbe refractometers can meet many of the requirements for routine measurements.
Single wavelengths and standard temperatures can be measured at extended ranges, as well as elevated temperatures, measurements of solid samples (films, plastic sheets).
Bestill nå
SPESIFIKASJONER
Modell NAR-2T NAR-3T NAR-4T NAR-1T LIQUID NAR-1T SOLID
Environmentally Preferable
Lyskilde LED-lamper LED LED-lamper LED-lamper LED-lamper
Temperaturmåling 5...120 °C 5...50 °C 5...50 °C 5...50 °C 5...50 °C
Temperatursensor ±0,2 °C ±0,2 °C ±0,2 °C ±0,2 °C ±0,2 °C
Temperatursensornøyaktighet ±0,2/0,5 °C ±0,2 °C ±0,2 °C ±0,2 °C ±0,2 °C