Cefazolin sodium salt
Leverandør: MP Biomedicals
Danger
Synonymer: Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
0215494680 0215494690 0215494691
ICNA0215494680EA 950 NOK
ICNA0215494680 ICNA0215494690 ICNA0215494691
Cefazolin sodium salt
Cefazolin sodium salt
Storage temperature.: +4 °C
Formel: C₁₄H₁₄N₈NaO₄S₃
MW: 476.48 g/mol
MDL nummer: MFCD00056883
CAS-nummer: 27164-46-1