Vaskemiddel, flytende, for automatisert rengjøring av glassutstyr, neodisher® FLA
Leverandør: Dr. Weigert
411230 411233 411226
148-3144EA 2375 NOK
148-3144 148-3222 148-3149
Vaskemiddel, flytende, for automatisert rengjøring av glassutstyr, neodisher® FLA
Rengjøringsløsninger Detergenter og overflatevasking
neodisher® LaboClean FLA.
  • Alkaline liquid concentrate for universal applications, free of phosphates, surfactants and oxidising agents
  • Components: 5 to 15% polycarboxylates
  • Dosage: 2 to 6 ml/L
NB: Most of the materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.