Multiparametermåler (pH/ISE/konduktivitet/DO), benk, inoLab® Multi 9620 IDS
Leverandør: WTW
1FD560 1FD56C
665-0476EA 44725 NOK
665-0476 665-0560
Multiparametermåler (pH/ISE/konduktivitet/DO), benk, inoLab® Multi 9620 IDS
Elektrokjemiske målere pH-meter
Digital high performance multi-parameter device with two galvanically isolated measuring channels for pH/mV, ISE (including incremental methods), conductivity and oxygen measurements. With comprehensive GLP functions.
  • Simultaneous measurement with two independent measuring channels
  • User-defined combinations of equal or different parameters
  • Continuous Measurement Control (CMC) for measuring in the optimum pH range
  • Quality Sensor Control (QSC) benchmarks the current condition of the special IDS electrodes
  • Digital processing and data transfer (GLP compliant)
More Product Information
Bestill nå
SPESIFIKASJONER
Modell inoLab® Multi 9620
Environmentally Preferable
pH - område Depending on sensor
pH - kalibrering 1- to 5-point
mV - område Depending on sensor
ISE kalibrering 2- to 7-point, adapter required
Konduktivitetsområde Depending on sensor
Konduktivitetskalibrering 1-point
DO konsentrasjon Depending on sensor
Oppløst oksygen - metning Depending on sensor
DO kalibrering 1-point
temperaturkompensering All except ORP
Display 16:9 colour graphic display
Strømforsyning Universal power supply (100 - 240 V, 50/60 Hz)
Datalogg - loggfunksjon Manual or time-controlled
Datalogg kapasitet Manual: 500 data sets; automatic: 10000 data sets
Utganger USB-A, Mini USB-B