Size exclusion and ligand exchange chromatography columns, SUGAR KS-802 and KS-801
Supplier: Resonac
F6378010 F6378020
554-2214EA 23075 NOK
554-2214 554-2215
Size exclusion and ligand exchange chromatography columns, SUGAR KS-802 and KS-801
Chromatography Columns
These columns separate mono and di-saccharides by combination of ligand exchange and size exclusion mode.
  • Counter ion: Na⁺
  • Can be used in tandem: KS-802 and KS-801
  • Only water is required for the analysis
  • Base material is styrene divinylbenzene copolymer
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. pressure 5,0 MPa (50 bar)
pH range 3 - 7
Temperature limit 20 - 85 °C
USP code L22, L58