Reversed phase chromatography columns, RSpak DM-614
Leverandør: Resonac
F7001002 F6700160
554-2916EA 13575 NOK
554-2916 554-2873
Reversed phase chromatography columns, RSpak DM-614
Kromatografikolonner
These columns are suitable for the analysis of amino acids and water-soluble vitamins.
  • Base material is polyhydroxymethacrylate
Bestill nå
SPESIFIKASJONER
Beskrivelse RSpak DM-614 RSpak DM-G 4A
Environmentally Preferable
Maks. trykk 2,0 MPa (20 bar)
pH område 2 - 10 2 - 10
Temperaturgrense 20 - 60 °C 20 - 60 °C
USP kode L39 L39