Reversed phase chromatography columns, RSpak DC-613
Leverandør: Resonac
F7001003 F6700170
554-2917EA 13575 NOK
554-2917 554-2874
Reversed phase chromatography columns, RSpak DC-613
Kromatografikolonner
These columns are used to separate by combination of ligand exchange and HILIC modes and can analyse sugars without removing sodium salts in the sample.
  • Counter ion: Na⁺
  • Base material is styrene divinylbenzene copolymer
Bestill nå
SPESIFIKASJONER
Beskrivelse RSpak DC-613 RSpak DC-G 4A
Environmentally Preferable
Maks. trykk 5,0 MPa (50 bar)
pH område 2 - 13 2 - 13
Temperaturgrense 40 - 85 °C 40 - 85 °C
USP kode L22, L58 L22, L58