Filtration capsules, AcroPak™ 20
Leverandør: Cytiva (Formerly Pall Lab)
12203 12200 12201 12202 12247
514-4097EA 2600 NOK
514-4097 514-4094 514-4095 514-4096 516-6945
Filtration capsules, AcroPak™ 20
Filter Membranfilterkapsler
AcroPak™ 20 capsule filters are designed to efficiently process up to 2 litres of chemical and biological fluids. These filters are available in 3 different membrane types.
Supor®
The Supor® membrane has high flow rates and throughputs, and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones. For sterile filtration of media and buffers.
• Built-in prefilter extends filter life when particulate-laden solutions such as serum-containing media are processed
• Sterilisation by gamma irradiation eliminates potential contamination by EtO residuals
More Product Information
Bestill nå
SPESIFIKASJONER
Type Fluorodyne® II Supor® Supor® EKV
Environmentally Preferable
Vanngjennomstrømning 26 ml/min (0,1 bar) 40 ml/min (0,1 bar)
Filtreringsområde 20 cm²
Inngang/utgang 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet 6,4 - 12,7 mm Ø stepped hose barb with female Luer slip interior and filling bell outlet 6,4 - 12,7 mm Ø stepped hose barbs with female slip Luer ID in the hose barb
Maks. temp omgivelser 60 °C 60 °C 60 °C
Maks. trykk 4,1 bar 4,1 bar 4,1 bar