7-Ethoxyresorufin ≥95%, solid
Leverandør: Cayman Chemical
Synonymer: Ethoxyresorufin, 7-Ethoxy-3H-phenoxazin-3-one, Resorufin ethyl ether
16122-10 16122-1
CAYM16122-10EA 6625 NOK
CAYM16122-10 CAYM16122-1
7-Ethoxyresorufin ≥95%, solid
7-Ethoxyresorufin
7-Ethoxyresorufin (7-ER) is a fluorometric substrate for and competitive inhibitor of cytochrome P450 1A1 (Km ranges from 0,11-0,23 µM; IC50 = 0,1 µM)
It has been used to study the metabolism of arachidonic acid and the mechanisms of vasodilation. 7-ER also has been shown to inhibit nitric oxide synthesis by uncoupling neuronal nitric oxide synthase (Ki = 0,76 µM)
Formel: C₁₄H₁₁NO₃
MW: 241.246 g/mol
MDL nummer: MFCD00037661
CAS-nummer: 5725-91-7