Crucibles, Conical, Flat Base, Aluminum Oxide
Leverandør: Thermo Fisher Scientific
33009.KF 33010.KF 33012.KF 33013.KF 33014.KF 33015.KF 33046.KF 33048.KF
33009.KFEA 1125 NOK
33009.KF 33010.KF 33012.KF 33013.KF 33014.KF 33015.KF 33046.KF 33048.KF
Crucibles, Conical, Flat Base, Aluminum Oxide
Digler
These crucibles are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • 1950 °C (max service temp in air)