Horizontal gel electrophoresis units, Axygen®
Leverandør: Corning
HGB-7 HGB-10 HGB-15 HGB-20
AXYGHGB-7EA 5500 NOK
AXYGHGB-7 AXYGHGB-10 AXYGHGB-15 AXYGHGB-20
Horizontal gel electrophoresis units, Axygen®
Elektroforesesystemer Horisontal gelelektroforesesystemer
Axygen® horizontal gel box is a high quality, full-featured unit designed for exceptional durability and efficiency. These gel boxes feature leak-free external gel casting, which allows the user to cast one gel outside of the box while another gel is running inside. Boxes from 7, 10, 15 and 20cm
  • Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
  • External gel casting to gel box allows for running gels while casting the next one
  • Ease of use – the Axygen contrast enhancer makes loading gels easier
  • Rugged injection molding is
More Product Information
Bestill nå
SPESIFIKASJONER
Modell HGB-7 HGB-10
Environmentally Preferable
Tankvolum 300 ml (buffer volume) 500 ml (buffer volume) 1500 ml (buffer volume)
Bredde 12.5 cm 17,5 cm 23 cm
Lengde 26.5 cm 39.5 cm