Neohesperidin, Reference Standards
Leverandør: USP
1458042 1458086
1458042.EA 10075 NOK
1458042. 1458086.
Neohesperidin, Reference Standards
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sertifiseringer: US Pharmacopea Reference Standard
Bestill nå
SPESIFIKASJONER
Beskrivelse Neohesperidin Neohesperidin Dihydrochalcone
Synonym(s) (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-oxo-3,4-dihydro-2H-chromen-7-yl 2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside 3,5-Dihydroxy-4-[3-(3-hydroxy-4-methoxyphenyl)propanoyl]phenyl 2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside
CAS-nr 13241-33-3 20702-77-6
Formel C₂₈H₃₄O₁₅