Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Leverandør: EUTECH
ELITEPH ELITEPHKIT ELITEPHSPEAR ELITEPHSPEARKI
662-2303EA 1800 NOK
662-2303 662-2280 662-2304 662-2282
Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
Elektrokjemiske målere pH-meter
These rugged and user-friendly testers feature a simultaneous display of pH and temperature and provide accurate measurements for water analysis. Available in a standard version or with a spear tip open pore sensor for direct measurements of solid or semi-solid samples such as cheese, meat, fruit, bread and more.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; one-push calibration
  • Easy to replace sensor
  • Lanyard provision for ‘on the go’ measurements
  • IP 67 waterproof rating
  • Standard version only: Leakproof cap with fill-line
More Product Information
Bestill nå
SPESIFIKASJONER
Modell Standard Standard Spear
Environmentally Preferable
pH - område −1,00...+15,00 −1,00...+15,00 −1,00...+15,00
pH - oppløsning 0,01 0,01 0,01
pH - nøyaktighet ±0,01 pH ±1 LSD ±0,01 pH ±1 LSD ±0,01 pH ±1 LSD
pH - kalibrering Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18) Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18) Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18)
Temperaturområde 0...+60,0 °C 0...+60,0 °C 0...+60,0 °C
Temperaturoppløsning 0,1 °C 0,1 °C 0,1 °C
Temperaturnøyakighet ±0,5 + 1 LSD (0...+50 °C)
±1,0 + 1 LSD (50...60 °C)
±0,5 + 1 LSD (0...+50)
±1,0 + 1 LSD (50...60)
-
temperaturkompensering Automatic (ATC) Automatic (ATC) Manual (MTC)
Display 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel) 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel) 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel)
Strømforsyning 4× AAA batteries 4× AAA batteries 4× AAA batteries
Samsvar IP 67 IP 67 IP 67
B×D×H 45×36×200 mm 45×36×200 mm 45×36×200 mm
Vekt 0,165 kg 0,165 kg 0,165 kg