Phosphatase assay kit, BIOMOL® Green
Leverandør: ENZO LIFE SCIENCES
BMLAK1110250 BMLAK1111000
ENZOBMLAK1110250EA 2375 NOK
ENZOBMLAK1110250 ENZOBMLAK1111000
Phosphatase assay kit, BIOMOL® Green
Assay Enzymanalyser
BIOMOL® Green provides a simple and convenient method for colorimetric phosphate quantitation. It is suitable for applications include assays for phospholipid phosphatases, tyrosyl-tRNA synthetase (coupled with pyrophosphatase) and a viral RNA triphosphatase. It is most widely applied to protein phosphatase assays.
  • Convenient one-step reagent, no mixing needed
  • Extremely stable with a long shelf-life, >6 months at +4 °C
  • Excellent sensitivity, ~50 pmol in 100 µl
  • Can be used in cuvette or microplate-based assays, making it ideal for high-throughput applications
Leveringsinformasjon: Kit contains Biomol® Green reagent and Phosphate standard.