Anti-DLK1 Mouse Monoclonal Antibody [clone: PF299-1]

Leverandør: ProSci Inc.
36-071
PRSI36-071EA 7225 NOK
PRSI36-071
Anti-DLK1 Mouse Monoclonal Antibody [clone: PF299-1]
Antistoffer
DLK1 (Protein delta homolog 1; Preadipocyte factor 1; Pref-1) is a regulator of adipocyte differentiation found in serum and urine. DLK1 might be a decoy ligand. Serum or plasma levels of soluble DLK1 affect negatively or positively adipogenesis and control mesenchymal cell fate. DLK1 is frequently upregulated in myelodysplastic syndrome (MDS) patients compared to non-leukemic individuals.

Type: Primary
Antigen: DLK1
Clonality: Monoclonal
Clone: PF299-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR