Discs for Qiagen Rotor-Gene™ Q real-time rotary analyser, RotoCycler™
Leverandør: Simport Scientific
T319-72D2 T319-100D1
SIMPT319-72D2EA 3100 NOK
SIMPT319-72D2 SIMPT319-100D1
Discs for Qiagen Rotor-Gene™ Q real-time rotary analyser, RotoCycler™
PCR-maskiner
The RotoCycler™ discs, made from PP, are specially designed to be used with the Qiagen Rotor-Gene™ Q real-time PCR cycler.
The discs are a one-piece “plate” equivalent, having vertically oriented wells compatible with automated reaction set-up using a robotic liquid handling system.
Two models are available:
72-well disc with 100 µl tubes (recommended reaction volume 20-25 µl) for use with the original Rotor-Disc® 72 Rotor and Rotor-Disc® 72 Locking Ring.
100-well disc with 30 µl tubes (recommended reaction volume 15-20 µl) for use with the original Rotor-Disc® 100 Rotor and Rotor-Disc® 100 Locking Ring.