Cell counting kit-8
Leverandør: ENZO LIFE SCIENCES
ALX850039KI01 ALX850039KI02
ENZOALX850039KI01EA 2950 NOK
ENZOALX850039KI01 ENZOALX850039KI02
Cell counting kit-8
Assay Cellulær analyse
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages: One-step, ready-to-use solution, no radioisotopes, no solubilization step, high sensitivity, correlates with the [3H]-thymidine incorporation assay. Can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results.
Bestillingsinformasjon: Quantity: 500 tests/5ml; 2500 tests/5 x 5ml. Contents: 5mM WST-8 solution, 0.2mM 1-methoxy-PMS, 150mM sodium chloride.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Størrelse 500 tests 5×500 tests