LDH Cytotoxicity WST Assay
Leverandør: ENZO LIFE SCIENCES
ENZKIT1570500 ENZKIT1572500
ENZOENZKIT1570500EA 3475 NOK
ENZOENZKIT1570500 ENZOENZKIT1572500
LDH Cytotoxicity WST Assay
Assay Cellulær analyse
Colorimetric kit for the determination of cytotoxicity through the measurement of lactate dehydrogenase activity.
  • Homogenous assay using living cells or non-homogenous assay protocol using cell culture supernatant
  • Stable working solution
The LDH cytotoxicity WST assay is a colorimetric assay kit used to determine cytotoxicity by measuring lactate dehydrogenase activity released from damaged cells.