Mouse recombinant IL33 (from E. coli)
Leverandør: ENZO LIFE SCIENCES
ALX5221013010 ALX522101C010
ENZOALX5221013010EA 9850 NOK
ENZOALX5221013010 ENZOALX522101C010
Mouse recombinant IL33 (from E. coli)
Proteiner
Produced in E. coli. Untagged mouse IL-33 (aa 109-266).
Bestill nå
SPESIFIKASJONER
Protein/peptide name IL33 IL33
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Arter Mouse Mouse
Kilde E. coli E. coli
Biologisk aktivitet Activates mouse and human ST2-dependent NF-κB pathway. Induces IL-6 production in P815 mastocytoma cells and in bone marrow derived mast cells. Activates mouse and human ST2-dependent NF-κB pathway. Induces IL-6 production in P815 mastocytoma cells and in bone marrow derived mast cells.
Renhet >95% (SDS-PAGE) >95% (SDS-PAGE)
Konsentrasjon 0,1 mg/ml after reconstitution 0,1 mg/ml after reconstitution
Formulering Lyophilised. Contains PBS. Lyophilised. Contains PBS.
Reconstitution instructions Reconstitute with 100 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum. Reconstitute with 100 µl sterile distilled water. Further dilutions should be made with medium containing 5% fetal calf serum.