Ionotropic glutamatergic ligand library, Screen-Well®
Supplier: ENZO LIFE SCIENCES
BML28170100 BML28170500
ENZOBML28170100EA 9175 NOK
ENZOBML28170100 ENZOBML28170500
Ionotropic glutamatergic ligand library, Screen-Well®
Cell Culture Supplements and Additives
The Screen-Well® ionotropic glutamatergic ligands plate contains 60 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The ionotropic glutamatergic ligands plate is 1 plate of the larger neurotransmitter library which contains over 680 ligands.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more