Bulk media for ion exchange chromatography columns, PL-SCX
Leverandør: VARIAN
PL1445-4102 PL1445-4103 PL1445-4702 PL1445-4703 PL1445-6102 PL1445-6103 PL1445-6702 PL1445-6703
VARIPL1445-4102EA 100175 NOK
VARIPL1445-4102 VARIPL1445-4103 VARIPL1445-4702 VARIPL1445-4703 VARIPL1445-6102 VARIPL1445-6103 VARIPL1445-6702 VARIPL1445-6703
Bulk media for ion exchange chromatography columns, PL-SCX
Kromatografiabsorbenter HPLC sorbenter
PL-SCX is a macroporous PS/DVB matrix with a very hydrophilic coating and strong cation exchange functionality. The manufacturing process is controlled to provide optimum density of strong cation-exchange moieties for analysing, separating, and purifying a wide range of biomolecules from small peptides to large proteins.
  • Reliable higher-resolution separations
  • Exceptional stability for long column lifetime
  • Chemical stability for the widest range of eluents and clean-up procedures for purification