Tinn ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Leverandør: Thermo Fisher Scientific
11459.FIEA 1200 NOK
11459.FI 11459.GH
Tinn ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Tinn
Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈4.6g/50x50mm Odour: Odourless MDL No.: MFCD00133862
Formel: Sn
MW: 118.71 g/mol
Kokepunkt: 2270
Smeltepunkt: 232
Tetthet: 7.31 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00133862
CAS-nummer: 7440-31-5
EINECS: 231-141-8
Merck Indeks: 12,09587