Tantalum ≥99.95% (metals basis), folie, Thickness: 2.0 mm (0.08 in)
Leverandør: Thermo Fisher Scientific
10357.FF 10357.FI 10357.GH
10357.FFEA 3025 NOK
10357.FF 10357.FI 10357.GH
Tantalum ≥99.95% (metals basis), folie, Thickness: 2.0 mm (0.08 in)
Tantalum
Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈20.8g/25x25mm Odour: Odourless MDL No.: MFCD00011252
Formel: Ta
MW: 180.95 g/mol
Kokepunkt: 5427
Smeltepunkt: 2996
Tetthet: 16.69 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011252
CAS-nummer: 7440-25-7
EINECS: 231-135-5
Merck Indeks: 12,09223